4-Fluoro-2,6-dimethylphenol ≥95%

Supplier: AOBChem USA

Warning

35356-25G 35356-5G
76931-642EA 569.71 USD
76931-642 76931-644
4-Fluoro-2,6-dimethylphenol ≥95%
4-Fluoro-2,6-dimethylphenol
Formula: C₈H₉FO
MW: 140.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972873
CAS Number: 2338-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR