5-Bromo-2,7-dimethyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

35302-1G 35302-25G 35302-500MG 35302-5G
76931-626EA 124.13 USD
76931-626 76931-628 76931-630 76931-632
5-Bromo-2,7-dimethyl-2H-indazole ≥97%
5-Bromo-2,7-dimethyl-2H-indazole
Formula: C₉H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12028637
CAS Number: 1146637-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR