4-Ethoxy-3,5-dimethylbenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

35219-1G 35219-25G 35219-500MG 35219-5G
76931-612EA 124.13 USD
76931-612 76931-614 76931-616 76931-618
4-Ethoxy-3,5-dimethylbenzaldehyde ≥97% research grade
4-Ethoxy-3,5-dimethylbenzaldehyde
Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06247333
CAS Number: 861376-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR