4-Bromo-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

35108-1G 35108-25G 35108-500MG 35108-5G
76931-596EA 346.92 USD
76931-596 76931-598 76931-600 76931-602
4-Bromo-3-fluoro-2-methylbenzoic acid ≥97%
4-Bromo-3-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18917158
CAS Number: 1349716-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR