4-Bromo-2-ethyl-6-methylaniline ≥95%

Supplier: AOBChem USA

Warning

35052-1G 35052-25G 35052-5G
76931-582EA 84.88 USD
76931-582 76931-584 76931-586
4-Bromo-2-ethyl-6-methylaniline ≥95%
4-Bromo-2-ethyl-6-methylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
Storage Temperature: Refrigerator
MDL Number: MFCD04093463
CAS Number: 70598-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR