4-Iodo-2-methoxypyridine-3-carbonitrile ≥95%

Supplier: AOBChem USA

Warning

34844-1G 34844-25G 34844-500MG 34844-5G
76931-540EA 153.84 USD
76931-540 76931-542 76931-544 76931-546
4-Iodo-2-methoxypyridine-3-carbonitrile ≥95%
4-Iodo-2-methoxypyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅IN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD14585335
CAS Number: 1206969-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR