4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

34812-1G 34812-500MG 34812-5G
76931-526EA 524.09 USD
76931-526 76931-528 76931-530
4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid ≥97%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695851
CAS Number: 2092502-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR