5-Bromo-2-fluoro-3-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
34798-1G
34798-25G
34798-500MG
34798-5G
76931-512EA
132.62
USD
76931-512
76931-514
76931-516
76931-518
5-Bromo-2-fluoro-3-methoxybenzoic acid ≥97%
5-Bromo-2-fluoro-3-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₃ MW: 249.034 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22570364 CAS Number: 1782260-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...