5-Bromo-2,4-difluorobiphenyl ≥97%

Supplier: AOBChem USA

Warning

34796-1G 34796-25G 34796-500MG 34796-5G
76931-498EA 263.11 USD
76931-498 76931-500 76931-502 76931-504
5-Bromo-2,4-difluorobiphenyl ≥97%
5-Bromo-2,4-difluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrF₂
Storage Temperature: Refrigerator
MDL Number: MFCD31696112
CAS Number: 2230481-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR