5-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34717-25G 34717-5G
76931-464EA 230.22 USD
76931-464 76931-466
5-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97%
5-Fluoro-2-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MW: 194.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236286
CAS Number: 238742-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR