4-Chloro-2-fluorobenzeneboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-fluorophenylboronic acid pinacol ester, 2-(4-Chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

34692-25G 34692-5G
76931-454EA 453.01 USD
76931-454 76931-456
4-Chloro-2-fluorobenzeneboronic acid pinacol ester ≥97% research grade
4-Chloro-2-fluorobenzeneboronic acid pinacol ester
Formula: C₁₂H₁₅BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD18729919
CAS Number: 765917-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR