4-Phenylthiophene-2-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

34676-25G 34676-5G
76931-442EA 268.41 USD
76931-442 76931-444
4-Phenylthiophene-2-carbaldehyde ≥95%
4-Phenylthiophene-2-carbaldehyde
Formula: C₁₁H₈OS
Storage Temperature: Refrigerator
MDL Number: MFCD03701610
CAS Number: 26170-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR