4-Chloro-4'-hydroxybiphenyl ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-ol, 4'-Chlorobiphenyl-4-ol

34587-1G 34587-25G 34587-5G
76931-432EA 70.02 USD
76931-432 76931-434 76931-436
4-Chloro-4'-hydroxybiphenyl ≥97%
4-Chloro-4'-hydroxybiphenyl
Formula: C₁₂H₉ClO
MW: 204.65 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00143298
CAS Number: 28034-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR