5-Benzyloxy-2-methylbenzoic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

34569-1G 34569-500MG 34569-5G
76931-420EA 598.35 USD
76931-420 76931-422 76931-424
5-Benzyloxy-2-methylbenzoic acid ethyl ester ≥97%
5-Benzyloxy-2-methylbenzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD31619502
CAS Number: 2221812-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR