4-Chloro-3-iodo-1H-indazole ≥95%
Supplier: AOBChem USA
|
Warning
|
34553-25G
34553-5G
76931-410EA
519.85
USD
76931-410
76931-412
4-Chloro-3-iodo-1H-indazole ≥95%
4-Chloro-3-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄ClIN₂ MW: 278.4800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781581 CAS Number: 518990-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...