4-Fluoro-2-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34401-1G 34401-500MG 34401-5G
76931-370EA 561.22 USD
76931-370 76931-372 76931-374
4-Fluoro-2-methoxy-3-methylbenzoic acid ≥97%
4-Fluoro-2-methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415174
CAS Number: 1301718-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR