4-Bromo-3,5-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

34318-25G 34318-5G
76931-362EA 495.45 USD
76931-362 76931-364
4-Bromo-3,5-difluorophenol ≥97%
4-Bromo-3,5-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD08062385
CAS Number: 130191-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR