4-Bromo-3-chloro-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

34107-1G 34107-25G 34107-500MG 34107-5G
76931-334EA 200.52 USD
76931-334 76931-336 76931-338 76931-340
4-Bromo-3-chloro-2-fluorophenylboronic acid pinacol ester ≥97%
4-Bromo-3-chloro-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BBrClFO₂
MW: 335.405 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619466
CAS Number: 2246613-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR