4-Methoxy-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33845-25G 33845-5G
76931-288EA 559.1 USD
76931-288 76931-290
4-Methoxy-3-(trifluoromethyl)benzyl alcohol ≥97%
4-Methoxy-3-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01631577
CAS Number: 261951-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR