4-Chloro-3-fluoro-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33822-1G 33822-500MG 33822-5G
76931-264EA 672.62 USD
76931-264 76931-266 76931-268
4-Chloro-3-fluoro-2-methylphenylboronic acid pinacol ester ≥97%
4-Chloro-3-fluoro-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClFO₂
MW: 270.535 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31561335
CAS Number: 1680200-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR