4-Bromo-3-(Methylsulfanyl)Benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33799-1G 33799-500MG 33799-5G
76931-240EA 635.48 USD
76931-240 76931-242 76931-244
4-Bromo-3-(Methylsulfanyl)Benzaldehyde ≥97%
4-Bromo-3-(Methylsulfanyl)Benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrOS
Storage Temperature: Ambient
MDL Number: MFCD29122065
CAS Number: 1289015-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR