4-Fluoro-2-isopropoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33428-1G 33428-25G 33428-500MG 33428-5G
76931-188EA 171.87 USD
76931-188 76931-190 76931-192 76931-194
4-Fluoro-2-isopropoxyphenylboronic acid pinacol ester ≥97%
4-Fluoro-2-isopropoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BFO₃
MW: 280.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039841
CAS Number: 1688656-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR