4-Chloro-3-(trifluoromethoxy)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33378-25G 33378-5G
76931-180EA 350.1 USD
76931-180 76931-182
4-Chloro-3-(trifluoromethoxy)benzyl alcohol ≥97%
4-Chloro-3-(trifluoromethoxy)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O₂
MW: 226.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660248
CAS Number: 886500-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR