5-Bromo-1,4-difluoro-2-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

33344-25G 33344-5G
76931-172EA 310.85 USD
76931-172 76931-174
5-Bromo-1,4-difluoro-2-isopropoxybenzene ≥95%
5-Bromo-1,4-difluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834048
CAS Number: 2121514-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR