4-Hydroxy-2,3-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

33338-1G 33338-25G 33338-500MG 33338-5G
76931-160EA 263.11 USD
76931-160 76931-162 76931-164 76931-166
4-Hydroxy-2,3-dimethylbenzaldehyde ≥95%
4-Hydroxy-2,3-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414657
CAS Number: 58380-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR