4-Fluoro-3-isopropoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33245-1G 33245-25G 33245-5G
76931-130EA 82.76 USD
76931-130 76931-132 76931-134
4-Fluoro-3-isopropoxybenzoic acid ≥95%
4-Fluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28356763
CAS Number: 869990-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR