4-Iodophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Iodobenzeneboronic acid pinacol ester, 2-(4-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

33030-25G 33030-5G
76931-112EA 381.93 USD
76931-112 76931-114
4-Iodophenylboronic acid pinacol ester ≥97%
4-Iodophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09953499
CAS Number: 73852-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR