3,5-Difluoro-4-chlorophenol ≥97%

Supplier: AOBChem USA

Warning

32934-1G 32934-25G 32934-5G
76931-076EA 66.84 USD
76931-076 76931-078 76931-080
3,5-Difluoro-4-chlorophenol ≥97%
3,5-Difluoro-4-chlorophenol
Formula: C₆H₃ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06658279
CAS Number: 2268-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR