4-Ethoxy-3,5-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32780-1G 32780-25G 32780-5G
76931-038EA 84.88 USD
76931-038 76931-040 76931-042
4-Ethoxy-3,5-difluorobenzyl alcohol ≥97%
Formula: C₉H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09258690
CAS Number: 1017779-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR