5-Bromo-3-methyl-2-(propan-2-yloxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32738-1G 32738-25G 32738-500MG 32738-5G
76931-020EA 260.99 USD
76931-020 76931-022 76931-024 76931-026
5-Bromo-3-methyl-2-(propan-2-yloxy)benzaldehyde ≥97%
5-Bromo-3-methyl-2-(propan-2-yloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26059136
CAS Number: 1564722-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR