5-Bromo-2-hydroxy-3-methylbenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

32728-25G 32728-5G
76931-008EA 573.95 USD
76931-008 76931-010
5-Bromo-2-hydroxy-3-methylbenzaldehyde ≥97% research grade
5-Bromo-2-hydroxy-3-methylbenzaldehyde
Formula: C₈H₇BrO₂
MW: 215.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01040324
CAS Number: 33172-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR