(3-Chloro-5-fluoro-4-isopropoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-3-fluoro-4-isopropoxyphenylboronic acid

32645-1G 32645-25G 32645-5G
76930-994EA 94.43 USD
76930-994 76930-996 76930-998
(3-Chloro-5-fluoro-4-isopropoxyphenyl)boronic acid ≥95%
(3-Chloro-5-fluoro-4-isopropoxyphenyl)boronic acid
Formula: C₉H₁₁BClFO₃
MW: 232.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20441800
CAS Number: 2096335-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR