4-Ethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

32577-1G 32577-25G 32577-500MG 32577-5G
76930-970EA 308.73 USD
76930-970 76930-972 76930-974 76930-976
4-Ethylbenzamide ≥97%
4-Ethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00017616
CAS Number: 33695-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR