5-Bromo-2-fluoro-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

32402-1G 32402-25G 32402-500MG 32402-5G
76930-930EA 124.13 USD
76930-930 76930-932 76930-934 76930-936
5-Bromo-2-fluoro-4-methylphenol ≥97%
5-Bromo-2-fluoro-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD12405415
CAS Number: 1111096-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR