4-Chloro-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32395-1G 32395-25G 32395-500MG 32395-5G
76930-914EA 153.84 USD
76930-914 76930-916 76930-918 76930-920
4-Chloro-2-fluoro-3-methylbenzoic acid ≥97%
4-Chloro-2-fluoro-3-methylbenzoic acid
Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17169808
CAS Number: 153556-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR