5-Chloro-2,4-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32334-1G 32334-25G 32334-500MG 32334-5G
76930-886EA 153.84 USD
76930-886 76930-888 76930-890 76930-892
5-Chloro-2,4-dimethoxyphenylboronic acid ≥97%
5-Chloro-2,4-dimethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₄
MW: 216.427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23380991
CAS Number: 1393536-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR