5-Bromo-N-ethylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

32244-25G 32244-5G
76930-878EA 198.39 USD
76930-878 76930-880
5-Bromo-N-ethylnicotinamide ≥97%
5-Bromo-N-ethylnicotinamide

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09953023
CAS Number: 173999-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR