2-(4-Chloro-2-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-fluoro-3-methoxyphenylboronic acid pinacol ester

32215-1G 32215-25G 32215-500MG 32215-5G
76930-864EA 145.35 USD
76930-864 76930-866 76930-868 76930-870
2-(4-Chloro-2-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Chloro-2-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₇BClFO₃
MW: 286.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22493590
CAS Number: 1126321-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR