4-Chloro-2-fluoro-3-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

32178-1G 32178-25G 32178-500MG 32178-5G
76930-840EA 142.17 USD
76930-840 76930-842 76930-844 76930-846
4-Chloro-2-fluoro-3-methoxybenzonitrile ≥97%
4-Chloro-2-fluoro-3-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFNO
MW: 185.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19687208
CAS Number: 1126320-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR