4-Chloro-2,3-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32173-1G 32173-25G 32173-500MG 32173-5G
76930-828EA 159.14 USD
76930-828 76930-830 76930-832 76930-834
4-Chloro-2,3-difluorobenzyl alcohol ≥97%
4-Chloro-2,3-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD28750500
CAS Number: 1805647-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR