4-Chloro-2,3-difluorophenol ≥95% research grade

Supplier: AOBChem USA

Warning

32171-1G 32171-25G 32171-500MG 32171-5G
76930-812EA 254.62 USD
76930-812 76930-814 76930-816 76930-818
4-Chloro-2,3-difluorophenol ≥95% research grade
4-Chloro-2,3-difluorophenol
Formula: C₆H₃ClF₂O
MW: 164.537 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415633
CAS Number: 1261634-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR