5-Bromo-2-(2,2,2-trifluoroethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-(2,2,2-trifluoro ethoxy)pyridine

32149-25G 32149-5G
76930-802EA 466.8 USD
76930-802 76930-804
5-Bromo-2-(2,2,2-trifluoroethoxy)pyridine ≥97%
5-Bromo-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374892
CAS Number: 126728-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR