5-Fluoro-2-iodobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

31967-1G 31967-25G 31967-500MG 31967-5G
76930-766EA 159.14 USD
76930-766 76930-768 76930-770 76930-772
5-Fluoro-2-iodobenzaldehyde ≥97% research grade
5-Fluoro-2-iodobenzaldehyde
Formula: C₇H₄FIO
Storage Temperature: Ambient
MDL Number: MFCD07782057
CAS Number: 877264-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR