(4-Ethoxy-3-ethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Ethoxy-3-ethylphenylboronic acid

31760-1G 31760-500MG 31760-5G
76930-732EA 623.81 USD
76930-732 76930-734 76930-736
(4-Ethoxy-3-ethylphenyl)boronic acid ≥97%
(4-Ethoxy-3-ethylphenyl)boronic acid
Formula: C₁₀H₁₅BO₃
MW: 194.0353 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30344675
CAS Number: 2121512-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR