5-Fluoro-4-picoline-3-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31552-1G 31552-500MG 31552-5G
76930-698EA 906.01 USD
76930-698 76930-700 76930-702
5-Fluoro-4-picoline-3-boronic acid pinacol ester ≥95%
5-Fluoro-4-picoline-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MW: 237.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18712609
CAS Number: 1356483-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR