Methyl-4-fluoro-2-(trifluoromethyl)benzenecarboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-fluoro-2-(trifluoromethyl)benzoate, 4-Fluoro-2-trifluoromethylbenzoic acid methyl ester

31291-1G 31291-25G 31291-5G
76930-660EA 106.09 USD
76930-660 76930-662 76930-664
Methyl-4-fluoro-2-(trifluoromethyl)benzenecarboxylate ≥97%
Methyl-4-fluoro-2-(trifluoromethyl)benzenecarboxylate
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203708
CAS Number: 220141-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR