5-Chloro-2-fluoro-4-isopropoxyphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
31204-1G
31204-25G
31204-500MG
31204-5G
76930-632EA
409.51
USD
76930-632
76930-634
76930-636
76930-638
5-Chloro-2-fluoro-4-isopropoxyphenylboronic acid pinacol ester ≥97%
5-Chloro-2-fluoro-4-isopropoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₁BClFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD29088278 CAS Number: 2121513-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...