5-Chloro-2-fluoro-4-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

31197-1G 31197-25G 31197-500MG 31197-5G
76930-618EA 317.21 USD
76930-618 76930-620 76930-622 76930-624
5-Chloro-2-fluoro-4-hydroxyphenylboronic acid ≥97%
5-Chloro-2-fluoro-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22414546
CAS Number: 2121512-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR