Ethyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

31154-1G 31154-25G 31154-500MG 31154-5G
76930-590EA 290.69 USD
76930-590 76930-592 76930-594 76930-596
Ethyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Ethyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formula: C₁₅H₂₀BFO₄
Storage Temperature: Refrigerator
CAS Number: 474709-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR