4-Chloro-2-methylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Chloro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chloro-2-methylphenylboronic acid pinacol ester

31118-25G 31118-5G
76930-558EA 279.02 USD
76930-558 76930-560
4-Chloro-2-methylphenylboronic acid, pinacol ester ≥97%
4-Chloro-2-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₂
MW: 252.545 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729895
CAS Number: 1030832-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR